Anti-CAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-1306R-A488
: Bioss
- Antibody type:Primary
- Antigen name:CAP, adenylate cyclase-associated protein 1 (yeast)
- Antigen symbol:CAP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10487
- Antigen synonyms:CAP|CAP1-PEN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:130-189/189
- Purification:Purified by Protein A
- Pk:100 µl
Directly regulates filament dynamics and has been implicated in a number of complex developmental and morphological processes, including mRNA localization and the establishment of cell polarity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CAP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat