Anti-AES Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-12392R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Amino-terminal Enhancer of Split
- Antigen symbol:AES
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:166
- Antigen synonyms:AES-1|AES-2|TLE5|Grg-5|ESP1|GRG5|GRG
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
TLE5 is a 197 amino acid nuclear protein that belongs to the TLE family. Expressed predominately in fetal brain, liver, lung, heart and kidney and in adult muscle, TLE5 functions as either a homooligomer or a heterooligomer with other TLE family members and, through this association, dominantly represses the expression of TLE genes. In addition, TLE5 can repress NFkB-regulated gene expression and is thought to play an important role in initiating and maintaining cell differentiation events. Two isoforms of TLE5 exist due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AES
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat