Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin carboxyl-terminal hydrolase isozyme L3
- Antigen symbol:UCHL3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Ubiquitin thioesterase L3|Ubiquitin thiolesterase L3|UCH L3|UCH-L3|UCHL3_HUMAN.|Ubiquitin thiolesterase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
UCH-L1 is a member of a gene family whose products hydrolyze small C-terminal adducts of ubiquitin to generate the ubiquitin monomer. Expression of UCH-L1 is highly specific to neurons and to cells of the diffuse neuroendocrine system and their tumors. UCH-L1 is expressed in brain neurons. Examination of specific brain regions reveals expression in all areas tested, particularly in the substantia nigra. UCH-L1 represents 1 to 2% of total soluble brain protein. Its occurrence in Lewy bodies and its function in the proteasome pathway make it a compelling candidate gene in Parkinson disease. The gene which encodes UCH-L1 maps to human chromosome 4p14. The 230 amino acid human UCH-L3 protein is 54% identical to that of UCH-L1. UCH-L3 is the predominant thiol protease and has high-affinity binding sites for ubiquitin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: UCHL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat