Anti-TXNDC11 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-11439R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Thioredoxin domain-containing protein 11
- Antigen symbol:TXNDC11
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:EF-hand-binding protein 1|Thioredoxin domain-containing protein 11|2810408E11Rik|EFP1|RGD1307538|TXNDC11|AI427833.|TXD11_HUMAN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
May act as a redox regulator involved in DUOX proteins folding. The interaction with DUOX1 and DUOX2 suggest that it belongs to a multiprotein complex constituting the thyroid H(2)O(2) generating system. It is however not sufficient to assist DUOX1 and DUOX2 in H(2)O(2) generation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TXNDC11
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat