Order Entry
Austria
ContactUsLinkComponent
Anti-HSP 40-4/HDJ2/DNAJA1 Rabbit Polyclonal Antibody (Cy3®)
Anti-HSP 40-4/HDJ2/DNAJA1 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-11277R-CY3
Supplier:  Bioss
Anti-HSP 40-4/HDJ2/DNAJA1 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-11277R-CY3
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HSP 40-4/HDJ2/DNAJA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

DnaJ-like proteins interact with HSP 70 molecular chaperones and function to facilitate protein folding and mitochondrial protein import. HSP 40-4, also known as HDJ2, is the human DnaJ homolog that functions as a co-chaperone with a cysteine-rich zinc finger domain. The cellular redox enzyme thioredoxin interacts with HSP 40-4, and oxidation and reduction reversibly regulate HSP 40-4 function in response to the changing redox states of the cell. The zinc finger domain of HSP 40-4 may act as a redox sensor of chaperone-mediated protein-folding machinery, since HSP 40-4 inactivation leads to the oxidation of cysteine thiols and a simultaneous release of coordinated zinc. Loss of the HSP 40-4 protein may be linked to severe defects in spermatogenesis that involve aberrant androgen signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSP 40-4/HDJ2/DNAJA1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat