Anti-NLK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-10420R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Nemo-like kinase
- Antigen symbol:NLK
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Nemo-like kinase (NLK) plays a role in cell fate determination and is required for differentiation of bone marrow stromal cells. It acts downstream of MAP3K7 and HIPK2 to negatively regulate the canonical Wnt/beta-catenin signaling pathway and the phosphorylation and destruction of the MYB transcription factor. It may suppress a wide range of transcription factors by phosphorylation of the coactivator, CREBBP. (referenced from swissprot)
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Nemo-like kinase
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat