Order Entry
Austria
ContactUsLinkComponent
Anti-HIF1A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-HIF1A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-0737R-A555
 :  Bioss
Anti-HIF1A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-0737R-A555
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor)
  • Antigen symbol:
    HIF1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3091
  • Antigen synonyms:
    PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    351-400/826
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Functions as a master transcriptional regulator of the adaptive response to hypoxia. Under hypoxic conditions, activates the transcription of over 4 genes, including erythropoietin, glucose transporters, glycolytic enzymes, vascular endothelial growth factor, HILPDA, and other genes whose protein products increase oxygen delivery or facilitate metabolic adaptation to hypoxia. Plays an essential role in embryonic vascularization, tumor angiogenesis and pathophysiology of ischemic disease. Binds to core DNA sequence 5'-[AG]CGTG-3' within the hypoxia response element (HRE) of target gene promoters. Activation requires recruitment of transcriptional coactivators such as CREBPB and EP3. Activity is enhanced by interaction with both, NCOA1 or NCOA2. Interaction with redox regulatory protein APEX seems to activate CTAD and potentiates activation by NCOA1 and CREBBP. Involved in the axonal distribution and transport of mitochondria in neurons during hypoxia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HIF1A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat