Anti-FOXE1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-0446R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box E1
- Antigen symbol:FOXE1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2304
- Antigen synonyms:FKHL15|forkhead (Drosophila)-like 15|forkhead box E1 (thyroid transcription factor 2)|HFKH4|FOXE1|drosophila|FOXE2|homolog-like 15|forkhead box E1|HFKL5|Forkhead|TITF2|TTF-2|thyroid transcription factor-2|forkhead box E2|TTF2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:130-180/373
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Probable transcription factor. Could be involved in thyroid gland organogenesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FOXE1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse