Anti-Neurokinin B Receptor Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-0166R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Neurokinin B Receptor
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:6870
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:220-245/465
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This is a receptor for the tachykinin neuropeptide neuromedin-K (neurokinin B). It is associated with G proteins that activate a phosphatidylinositol-calcium second messenger system. The rank order of affinity of this receptor to tachykinins is: neuromedin-K >substance K >substance P.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Neurokinin B receptor
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat