ContactUsLinkComponent
Digital camera, AxioCam HR
Digital camera, AxioCam HR
Catalog # ZEIS4265109901000
Supplier:  ZEISS
undefined
Digital camera, AxioCam HR
Catalog # ZEIS4265109901000
Supplier:  ZEISS
Specifications
  • Model:
    AxioCam HRc
  • Resolution:
    1,4 MP
  • Sensor type:
    CCD
  • Sensor size:
    2/3"
  • Description:
    Digital camera
  • Seq:
    999
  • Pixel size:
    6,45×6,45 µm
  • Interfaces:
    FireWire IEEE 1394a
  • Sensor:
    Sony ICX 285 Colour CCD
  • Pk:
    1
Specifications
About this item
High resolution camera for complex applications in pathology, developmental biology and material science.
  • No rotating fans: Camera works without any vibrations
  • Very small dimensions and no external control box
  • Large CCD sensor ensures perfect colour accuracy
  • High performance at low light intensities
The AxioCam HR can be connected to any Carl Zeiss microscope that has a phototube or TV output.