ContactUsLinkComponent
WiFi temperature and humidity data loggers, new Saveris 2
WiFi temperature and humidity data loggers, new Saveris 2
Supplier:  Testo
Certificates
undefined
WiFi temperature and humidity data loggers, new Saveris 2
Supplier:  Testo
Specifications
  • Catalog No:
  • 620-2699
  • 620-2698
  • 620-2697
  • 620-2696
  • 620-2695
  • Model:
  • testo Saveris 2 -H2
  • testo Saveris 2 -H1
  • testo Saveris 2 -T3
  • testo Saveris 2 -T2
  • testo Saveris 2 -T1
  • Sensor:
  • NTC sensor
  • NTC int.
  • TC type K
    TC type J
    TC type T
  • NTC sensor
  • NTC int.
  • Alarm:
  • Time-controlled alarm, alarm delay
  • Time-controlled alarm, alarm delay
  • Time-controlled alarm, alarm delay
  • Time-controlled alarm, alarm delay
  • Time-controlled alarm, alarm delay
  • Measurement time:
  • 5 s - 24 hrs
  • 5 s - 24 hrs
  • 5 s - 24 hrs
  • 5 s - 24 hrs
  • 5 s - 24 hrs
  • Interface:
  • WiFi
  • WiFi
  • WiFi
  • WiFi
  • WiFi
  • Protection class:
  • IP 54
  • IP 40
  • IP 54
  • IP 65
  • IP 65
  • Battery life:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • Recording intervals:
  • 1 min - 24 hrs
  • 1 min - 24 hrs
  • 1 min - 24 hrs
  • 1 min - 24 hrs
  • 1 min - 24 hrs
  • Weight:
  • 240 g
  • 240 g
  • 240 g
  • 240 g
  • 240 g
  • W×D×H:
  • 95×75×30,5 mm
  • 95×75×30,5 mm
  • 95×75×30,5 mm
  • 95×75×30,5 mm
  • 95×75×30,5 mm
  • Data memory - No. of readings:
  • 2×10000
  • 2×10000
  • 2×10000
  • 2×10000
  • 10000
  • Batteries:
  • 4×AA batteries
  • 4×AA batteries
  • 4×AA batteries
  • 4×AA batteries
  • 4×AA batteries
Specifications
Probes
Product Information
Frequently Bought Together