ContactUsLinkComponent
J.T.Baker® BAKERBOND spe™ C18-SDB-1, SPE Columns for Special Applications
J.T.Baker® BAKERBOND spe™ C18-SDB-1, SPE Columns for Special Applications
Catalog # 7650-07
Supplier:  Avantor
J.T.Baker® BAKERBOND spe™ C18-SDB-1, SPE Columns for Special Applications
Catalog # 7650-07
Supplier:  Avantor
Specifications
  • Capacity:
    6 ml
  • Description:
    Column with 500 mg C18 (top) and 200 mg SDB-1, separated by a PE frit
  • Bed weight:
    700 mg
  • Pk:
    30
Specifications
About this item
These SPE columns are designed for the extraction of pesticides from water.