ContactUsLinkComponent
Trace element solution 2, MP Biomedicals
Trace element solution 2, MP Biomedicals
Catalog # ICNA091676649
Supplier:  MP Biomedicals
CAS Number:  
Trace element solution 2, MP Biomedicals
Catalog # ICNA091676649
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    Trace Element Solution 2
Specifications
About this item
Trace Element Solution 2 is a 1000X sterile filtered solution, suitable for use with serum-free media; ideal for plant cell culture.
Trace Element Solution 2 is added aseptically to media as a supplement. Prior to use of the media it may be necessary to adjust pH and sterile filter (to reduce contamination).