ContactUsLinkComponent
VWR® Beakers, Low Form, PMP (TPX)
VWR® Beakers, Low Form, PMP (TPX)
Catalog #
Supplier:  VWR Collection
   Sustainable Options Available
Supplier #:
undefined
VWR® Beakers, Low Form, PMP (TPX)
Catalog #
Supplier:  VWR Collection
   Sustainable Options Available
Supplier #:
Select an Option
Select an Option
Select an Option
Select an Option
Select an Option
Select an Option
Select an Option
341,0 g
Specifications
  • Catalog No:
  • 213-0485
  • 213-0484
  • 213-0487
  • 213-0486
  • 213-0482
  • 213-0510
  • 213-0483
  • 213-0488
  • Environmentally Preferable:
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
Specifications
About this item
Made of PMP (TPX), crystal-clear.
  • Excellent chemical resistance
  • Printed graduations
  • Withstand temperatures of 0 to 121 °C when used continuously (up to 170 °C for short periods)
  • Resin meets US FDA 21 CFR requirements
Frequently Bought Together