0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Scoops
Weighing scoop with knob
Weighing scoop with knob
Weighing scoop with knob
611-1252
:
USBECK Laborgeräte
Weighing scoop with knob
611-1252
:
USBECK Laborgeräte
Width:
30 mm
Length:
200 mm
Pk:
1
Stainless steel.
Corrosion resistant
Easy to clean
For a variety of uses