ContactUsLinkComponent
J.T.Baker® BAKERBOND spe™ PAH Soil, SPE Columns for Special Applications
J.T.Baker® BAKERBOND spe™ PAH Soil, SPE Columns for Special Applications
Catalog # 7518-08
Supplier:  Avantor
J.T.Baker® BAKERBOND spe™ PAH Soil, SPE Columns for Special Applications
Catalog # 7518-08
Supplier:  Avantor
Specifications
  • Capacity:
    6 ml
  • Description:
    Column with 500 mg cyano (top) and 1000 mg silica gel, separated by a PE frit
  • Bed weight:
    1500 mg
  • Phase:
    PAH SOIL
  • Pk:
    30
Specifications
About this item
This solid phase extraction column is designed for the extraction and clean-up of Poly Aromatic Hydrocarbons (PAH) from soil, including the 16 priority pollutant EPA PAH’s and for several applications using different SPE columns.