Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
 
Zinc chloride 97.0-100.5%, granular USP, J.T.Baker®
  4320-07
 :  Avantor
 :  
undefined
 
 
 
 
Zinc chloride 97.0-100.5%, granular USP, J.T.Baker®
  4320-07
 :  Avantor
 :  

 

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

 

 

  • GMP Manufactured Product:
  • Meets U.S.P Requirements:
  • Store at 2 to 25 °C:
  • Preserve in Tight Containers:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    = 0.03 %
  • Identification:
    Passes Test
  • Oxychloride:
    Passes Test
  • Sulfate (SO₄):
    ≤0.03 %
  • Alkalies and Alkaline Earths:
    ≤1.0 %
  • Ammonium Salts:
    Passes Test
  • Lead (Pb):
    ≤0.005 %
  • Assay (ZnCl₂):
    97.0 - 100.5 %

 

 

 :
 :136.3 g/mol
 :13,10185
 :732 °C (1013 hPa)
 :283 °C
 :2.91 g/cm³ (20 °C)
 :Ambient
 :231-592-0
 :2331
 :8,III
 :GHS05
 :GHS07
 :GHS09
 :GHS08
 :7646-85-7
 :ZnCl₂
 :MFCD00011295