Orders LinkContactUsLinkComponent
Pipe Systems, FlexTube and FlexTubePro, SymLine®
Pipe Systems, FlexTube and FlexTubePro, SymLine®
Supplier:   S.C.A.T.
Pipe Systems, FlexTube and FlexTubePro, SymLine®
Supplier:  S.C.A.T.
Certificates
Pipe Systems, FlexTube and FlexTubePro, SymLine®
Pipe Systems, FlexTube and FlexTubePro, SymLine®
8 Options Available Below
Product Details & Documents
These flexible tubes made from PTFE can be used in safety cabinets observing the specified maximum dimension for drillings. They are the ideal solution for installation in confined spaces since they can be placed easily into clearances and provide the flexibility required in safety cabinets with pull-out containers.
  • Made of PTFE for best possible chemical resistance
  • Adjustable and easy to install; shorten tube as required, then mount the optimized screw connection and sealing
  • Electrostatic conductive <106 (max) Ω
  • 16,9 or 19,8 mm Ext.Ø
  • Available in 4 different standard lengths: 600, 1200, 1500, and 2500 mm
FlexTube with inner wall spiral or or FlexTubePro Inner wall smooth. The two most flexible and adaptable tubing systems for the safe disposal of liquid HPLC waste. Flexible, expandable, chemically resistant PTFE tubing with a large bending radius. SymLine® FlexTube Pro with unruffled inner wall prevents waste and residues in tubes.
Product Family Options
FlexTube
Product Information
  • Tubing ext.Ø
    Length
    Availability
    Price
  • SCTV106569
    SymLine® FlexTube, GL 25 (f), electrostatic conductive PTFE
    SymLine® FlexTube, GL 25 (f), electrostatic conductive PTFE
    19,8 mm
    600 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,45 €
    87,45 €
    1 * 1 ST
       87,45 €
  • SCTV106568
    SymLine® FlexTube, GL 25 (f), electrostatic conductive PTFE
    SymLine® FlexTube, GL 25 (f), electrostatic conductive PTFE
    19,8 mm
    1000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 97,04 €
    97,04 €
    1 * 1 ST
       97,04 €
  • SCTV106567
    SymLine® FlexTube, GL 25 (f), electrostatic conductive PTFE
    SymLine® FlexTube, GL 25 (f), electrostatic conductive PTFE
    19,8 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 108,98 €
    108,98 €
    1 * 1 ST
       108,98 €
  • SCTV106566
    SymLine® FlexTube, GL 25 (f), electrostatic conductive PTFE
    SymLine® FlexTube, GL 25 (f), electrostatic conductive PTFE
    19,8 mm
    2500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 133,00 €
    133,00 €
    1 * 1 ST
       133,00 €
FlexTube Pro
Product Information
  • Tubing ext.Ø
    Length
    Availability
    Price
  • SCTV106575
    SymLine® FlexTube Pro, GL 25 (f), electrostatic conductive PTFE
    SymLine® FlexTube Pro, GL 25 (f), electrostatic conductive PTFE
    16,9 mm
    600 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,98 €
    125,98 €
    1 * 1 ST
       125,98 €
  • SCTV106574
    SymLine® FlexTube Pro, GL 25 (f), electrostatic conductive PTFE
    SymLine® FlexTube Pro, GL 25 (f), electrostatic conductive PTFE
    16,9 mm
    1000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 169,05 €
    169,05 €
    1 * 1 ST
       169,05 €
  • SCTV106573
    SymLine® FlexTube Pro, GL 25 (f), electrostatic conductive PTFE
    SymLine® FlexTube Pro, GL 25 (f), electrostatic conductive PTFE
    16,9 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,94 €
    223,94 €
    1 * 1 ST
       223,94 €
  • SCTV106572
    SymLine® FlexTube Pro, GL 25 (f), electrostatic conductive PTFE
    SymLine® FlexTube Pro, GL 25 (f), electrostatic conductive PTFE
    16,9 mm
    2500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 332,84 €
    332,84 €
    1 * 1 ST
       332,84 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Tube holder for Symline FlexTube Pro, suitable for all cabinets
    Tube holder for Symline FlexTube Pro, suitable for all cabinets
    SCTV106558
    Log in for availability
    1 * 1 ST - 16,38 €
Product Information
  • Catalog #
    Availability
    Price
  • Tube holder for Symline FlexTube, suitable for all cabinets
    Tube holder for Symline FlexTube, suitable for all cabinets
    SCTV106562
    Log in for availability
    1 * 1 ST - 16,38 €
Product Information
  • Catalog #
    Availability
    Price
  • Screw GL25 for FlexTube, set of 2 pcs.
    Screw GL25 for FlexTube, set of 2 pcs.
    SCTV106579
    Log in for availability
    1 * 1 ST - 64,28 €
Product Information
  • Catalog #
    Availability
    Price
  • Screw GL25 for FlexTube Pro, set of 2 pcs.
    Screw GL25 for FlexTube Pro, set of 2 pcs.
    SCTV106591
    Log in for availability
    1 * 1 ST - 66,14 €