ContactUsLinkComponent
VWR®, Measuring Beakers with Handle, PP
VWR®, Measuring Beakers with Handle, PP
Catalog # 213-0501
Supplier:  VWR Collection
   Sustainable Options Available
Supplier #:
Sustainable Packaging
undefined
VWR®, Measuring Beakers with Handle, PP
Catalog # 213-0501
Supplier:  VWR Collection
   Sustainable Options Available
Supplier #:
  Sustainable Packaging
Log in for availability
Select an Option
206,5 mm
Select an Option
100 ml
Select an Option
2000 ml
Select an Option
192,0 mm
Select an Option
05
Select an Option
1
Specifications
  • Catalog No:
  • 213-0501
  • 213-0497
  • 213-0499
  • 213-0498
  • 213-0500
  • 213-0503
  • 213-0502
  • Environmentally Preferable:
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
Specifications
About this item
Made of PP, translucent.
  • With printed graduations
  • With handle for better usage
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements
Frequently Bought Together