ContactUsLinkComponent
VWR®, Beakers, Low Form, PP
VWR®, Beakers, Low Form, PP
Catalog # 213-0495
Supplier:  VWR Collection
   Sustainable Options Available
Supplier #:
Sustainable Packaging
undefined
VWR®, Beakers, Low Form, PP
Catalog # 213-0495
Supplier:  VWR Collection
   Sustainable Options Available
Supplier #:
  Sustainable Packaging
Log in for availability
Select an Option
3000 ml
Select an Option
08
Select an Option
1
Specifications
  • Catalog No:
  • 213-0495
  • 213-0494
  • 213-0496
  • 213-0493
  • 213-0492
  • 213-0491
  • 213-0511
  • 213-0490
  • 213-0489
  • Environmentally Preferable:
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
Specifications
About this item
PP, translucent.
  • Printed graduations
  • Can be stacked when empty to save space while storing
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements
Ideal for general laboratory use. With spout and exceptionally 'easy to read' printed graduations.
Frequently Bought Together