Water with 0,1% (v/v) formic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Catalog # 1.59013.2500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
Boiling Pt:100 °C (1013 hPa)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
:64-18-6