Order Entry
Austria
ContactUsLinkComponent
Spatula, 'V' shape
Spatula, 'V' shape
Catalog # SAMP1095S-150
Supplier:  Sampling Systems
undefined
Spatula, 'V' shape
Catalog # SAMP1095S-150
Supplier:  Sampling Systems

Specifications

  • Length:
    230 mm
  • Blade width:
    35 mm
  • Pk:
    1
  • Blade length:
    145 mm

Specifications

About this item

This V spatula made from stainless steel 316 is ideal for surface sampling of powders and core sampling of viscous material like cheese, pastes and gels.

  • Polished finish
  • Fully welded, crevice free construction
  • ‘V’ shape allows easy discharge of the powders into containers or bags