ContactUsLinkComponent
Replacement beakers for Goldfisch fat extractor
Replacement beakers for Goldfisch fat extractor
Catalog # LBCP3505300
Supplier:  LABCONCO
undefined
Replacement beakers for Goldfisch fat extractor
Catalog # LBCP3505300
Supplier:  LABCONCO
Specifications
  • Capacity:
    100 ml
  • Description:
    Beakers for Goldfisch fat extractor
  • Height:
    83 mm
  • Pk:
    6
Specifications
About this item
Beakers specifically designed for the Goldfisch Fat Extractor. Thick-walled borosilicate glass with flanged and ground tops to facilitate connection to the condenser.