ContactUsLinkComponent
Inert gas sampling syringe valve
Inert gas sampling syringe valve
Catalog # HAMI86580
Supplier:  HAMILTON BONADUZ
undefined
Inert gas sampling syringe valve
Catalog # HAMI86580
Supplier:  HAMILTON BONADUZ
Specifications
  • Description:
    Inert gas sampling syringe valve
  • Pk:
    1
Specifications
About this item
This MTB sampling valve is a lever-actuated on/off valve used for storing and transporting samples in any Gastight® PTFE Luer lock (TLL) or Luer tip (LT) syringe.
  • Pressure tested to 100 psig
  • 0,99 mm inner bore
Convert any TLL or LT syringe into a SampleLock™ syringe using this simple accessory. The syringe attaches to the female Luer lock connection leaving the male Luer open for connection to any Kel-F® hub.