ContactUsLinkComponent
Software, EuroOsmo® 7400
Software, EuroOsmo® 7400
Catalog # KNAUA3705
Supplier:  KNAUER
Software, EuroOsmo® 7400
Catalog # KNAUA3705
Supplier:  KNAUER
Specifications
  • Description:
    EuroOsmo® 7400 software, for K-7400 and K-7400S osmometers
  • Pk:
    1
Specifications
About this item
EuroOsmo® 7400 is a user friendly software developed for the freezing point osmometers K-7400 and K-7400S.
  • Data can optionally be exported into xls or txt files
It allows control of the instruments and recording of the measured data. The software automatically plots the temperature curve for each measurement and calibration and allows saving of the measured values. As with the instrument itself, the user can choose between freezing point depression or osmolality to be displayed also in the software. The data can optionally be exported into xls or txt files for further analysis. For archival storage a detailed analysis report can additionally be printed.