Order Entry
Austria
ContactUsLinkComponent
Dynasore ≥98% (by HPLC), Millipore®
Dynasore ≥98% (by HPLC), Millipore®
Catalog # 324410-10
CAS Number:  
Dynasore ≥98% (by HPLC), Millipore®
Catalog # 324410-10
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :322.32 g/mol
Storage Temperature:Refrigerator
CAS Number:304448-55-3
Formula:C₁₈H₁₄N₂O₄
MDL Number:MFCD00292551