Orders LinkContactUsLinkComponent
Reducing solution for ORP electrodes
Reducing solution for ORP electrodes
Reducing solution for ORP electrodes, 500 ml bottle, 1
Catalog # HACH2965349
Supplier:   Hach
Reducing solution for ORP electrodes
Catalog # HACH2965349
Supplier:  Hach
CAS Number:  
Reducing solution for ORP electrodes
Reducing solution for ORP electrodes
Reducing solution for ORP electrodes, 500 ml bottle, 1
CAS Number:  
Price
50,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Used to condition ORP electrode surfaces for use in reducing sample matrices.
Documents
Specifications
Specifications
  • Pk:
    1
  • Description:
    Reducing solution for ORP electrodes, 500 ml bottle
Specifications