ContactUsLinkComponent
Alkaline detergents, liquid concentrate, LaboClean FM, neodisher®
Alkaline detergents, liquid concentrate, LaboClean FM, neodisher®
Catalog # 148-0159
Supplier:  Dr. Weigert
undefined
Alkaline detergents, liquid concentrate, LaboClean FM, neodisher®
Catalog # 148-0159
Supplier:  Dr. Weigert
Specifications
  • Description:
    LaboClean FM, Alkaline detergent
  • Pk:
    5 L
Specifications
About this item
LaboClean FM is an alkaline, surfactant-containing detergent with intensive cleaning action. Ideal for the removal of oil, fat, resin, paraffin and organic dyes.
  • High emulsifying and dispersion action
  • High soil carrying ability
  • Free of phosphates, silicates and oxidising agents
Suitable for alkali-compatible utensils made of glass, stainless steel, ceramic and plastics. It is not suitable for utensils made of aluminium and nonanodised aluminium.