Anti-COL4A6 Rabbit Polyclonal Antibody [clone: G1223]
Catalog # BWRLBS9130
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:COL4A6
- Clonality:Polyclonal
- Clone:G1223
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 1190-1240 of Human COL4A6.
COL4A6 (G1223) pAb detects endogenous levels of COL4A6 protein.
Type: Primary
Antigen: COL4A6
Clonality: Polyclonal
Clone: G1223
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human