Anti-p-PKC zeta Rabbit Polyclonal Antibody [clone: T410]
BWRLBS4156
- Antibody type:Primary
- Antigen symbol:p-PKC zeta
- Clonality:Polyclonal
- Clone:T410
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human PKC zeta around the phosphorylation site of Threonine 410.
p-PKC ? (T410) pAb detects endogenous levels of PKC ? protein only when phosphorylated at Thr410.
Type: Primary
Antigen: p-PKC zeta
Clonality: Polyclonal
Clone: T410
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat