Anti-IRS-1 Rabbit Polyclonal Antibody [clone: P618]
BWRLBS5582
- Antibody type:Primary
- Antigen symbol:IRS-1
- Clonality:Polyclonal
- Clone:P618
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 600-650 of Human IRS-1.
IRS-1 (P618) pAb detects endogenous levels of IRS-1 protein.
Type: Primary
Antigen: IRS-1
Clonality: Polyclonal
Clone: P618
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat