Anti-p-c-Met Rabbit Polyclonal Antibody [clone: Y1234]
Catalog # BWRLBS4752
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-c-Met
- Clonality:Polyclonal
- Clone:Y1234
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human c-Met around the phosphorylation site of Tyrosine 1234.
p-c-Met (Y1234) pAb detects endogenous levels of c-Met protein only when phosphorylated at Tyr1234.
Type: Primary
Antigen: p-c-Met
Clonality: Polyclonal
Clone: Y1234
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat