Anti-p-eIF4E Rabbit Polyclonal Antibody [clone: S209]
Catalog # BWRLBS4748
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-eIF4E
- Clonality:Polyclonal
- Clone:S209
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human eIF4E around the phosphorylation site of Serine 209.
p-eIF4E (S209) pAb detects endogenous levels of eIF4E protein only when phosphorylated at Ser209.
Type: Primary
Antigen: p-eIF4E
Clonality: Polyclonal
Clone: S209
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat