Order Entry
Austria
ContactUsLinkComponent
Anti-p-JNK1/2/3 Rabbit Polyclonal Antibody [clone: T183/Y185]
Anti-p-JNK1/2/3 Rabbit Polyclonal Antibody [clone: T183/Y185]
Catalog # BWRLBS4322
Anti-p-JNK1/2/3 Rabbit Polyclonal Antibody [clone: T183/Y185]
Catalog # BWRLBS4322

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-JNK1/2/3
  • Clonality:
    Polyclonal
  • Clone:
    T183/Y185
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human JNK1 around the phosphorylation site of Threonine 183 and Tyrosine 185.

P-JNK1/2/3 (T183/Y185) pAb detects endogenous levels of JNK1/2/3 protein only when phosphorylated at Thr183 and Thy185.

Type: Primary
Antigen: p-JNK1/2/3
Clonality: Polyclonal
Clone: T183/Y185
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat