Anti-Pmp24 Rabbit Polyclonal Antibody [clone: L27]
BWRLBS3366
- Antibody type:Primary
- Antigen symbol:Pmp24
- Clonality:Polyclonal
- Clone:L27
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminual of Human Pmp24.
Pmp24 (L27) pAb detects endogenous levels of Pmp24 protein.
Type: Primary
Antigen: Pmp24
Clonality: Polyclonal
Clone: L27
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human