Anti-CYB5R3 Rabbit Polyclonal Antibody [clone: I169]
BWRLBS3337
- Antibody type:Primary
- Antigen symbol:CYB5R3
- Clonality:Polyclonal
- Clone:I169
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 141-190 of Human CYB5R3.
CYB5R3 (I169) pAb detects endogenous levels of CYB5R3 protein.
Type: Primary
Antigen: CYB5R3
Clonality: Polyclonal
Clone: I169
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse