Anti-Golgin 45 Rabbit Polyclonal Antibody [clone: D22]
Catalog # BWRLBS2672
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Golgin 45
- Clonality:Polyclonal
- Clone:D22
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminual of Human Golgin 45.
Golgin 45 (D22) pAb detects endogenous levels of Golgin 45 protein.
Type: Primary
Antigen: Golgin 45
Clonality: Polyclonal
Clone: D22
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse