Anti-SMG7 Rabbit Polyclonal Antibody [clone: K548]
Catalog # BWRLBS2112
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:SMG7 nonsense mediated mRNA decay factor
- Clonality:Polyclonal
- Clone:K548
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 512-564 of Human SMG7.
SMG7 (K548) pAb detects endogenous levels of SMG7 protein.
Type: Primary
Antigen: SMG7
Clonality: Polyclonal
Clone: K548
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human