Anti-PP2A-B56-delta Rabbit Polyclonal Antibody [clone: E574]
Catalog # BWRLBS3942
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PP2A-B56- delta
- Clonality:Polyclonal
- Clone:E574
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 540-592 of Human PP2A-B56-delta.
PP2A-B56-? (E574) pAb detects endogenous levels of PP2A-B56-? protein.
Type: Primary
Antigen: PP2A-B56-delta
Clonality: Polyclonal
Clone: E574
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat