Order Entry
Austria
ContactUsLinkComponent
Anti-SERCA1 Rabbit Polyclonal Antibody [clone: D580]
Anti-SERCA1 Rabbit Polyclonal Antibody [clone: D580]
  BWRLBS3934
Anti-SERCA1 Rabbit Polyclonal Antibody [clone: D580]
  BWRLBS3934

 

  • Antibody type:
    Primary
  • Antigen name:
    Sarcoplasmic or Endoplasmic Reticulum Calcium ATPase 1
  • Antigen symbol:
    SERCA1
  • Clonality:
    Polyclonal
  • Clone:
    D580
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 551-600 of Human SERCA1.

SERCA1 (D580) pAb detects endogenous levels of SERCA1 protein.

Type: Primary
Antigen: SERCA1
Clonality: Polyclonal
Clone: D580
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat