Anti-E-cadherin Rabbit Polyclonal Antibody [clone: G2]
Catalog # BWRLBS1097
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:E-cadherin
- Clonality:Polyclonal
- Clone:G2
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminus of Human E-cadherin.
E-cadherin(G2) pAb detects endogenous levels of E-cadherin protein.
Type: Primary
Antigen: E-cadherin
Clonality: Polyclonal
Clone: G2
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat