Order Entry
Austria
ContactUsLinkComponent
 
Contact plate, violet red bile glucose (VRBG) agar
Catalog # APPC433745.0922
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Contact plate, violet red bile glucose (VRBG) agar
Catalog # APPC433745.0922
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    30
  • Description:
    VRBG (Violet red bile glucose) agar

Specifications

About this item

Violet red bile glucose agar (VRBG) is used as a culture media for microbiology.

  • pH: 7,4 ±0,2
  • Incubate at 30 to 35 °C for 24 to 48 hours
  • Storage in a cool and dry place

It contains bile salts mixture (1,5 g/L), crystal violet (0,002 g/L), neutral red (0,03 g/L), D(+)-glucose (10,0 g/L), yeast extract (3,0 g/L), gelatin peptone (7,0 g/L), sodium chloride (5,0 g/L) and agar (20,5 g/L).

Certifications: Meets Ph. Eur. and ISO 21528 standard specifications.