Order Entry
Austria
Orders LinkContactUsLinkComponent
Multi-parameter meter (pH/mV/DO), bench, Orion Star™ A216

Multi-parameter meter (pH/mV/DO), bench, Orion Star™ A216

Supplier:   Thermo Orion
Multi-parameter meter (pH/mV/DO), bench, Orion Star™ A216
Supplier:  Thermo Orion
Certificates
Multi-parameter meter (pH/mV/DO), bench, Orion Star™ A216

2 Options Available Below

Product Details & Documents

This benchtop meter is ideal for a wide range of applications and advanced pH and dissolved oxygen analysis in the lab. It offers two measuring channels allowing users to measure pH and dissolved oxygen simultaneously or view each channel separately. Can be used with RDO optical or polarographic DO probes.

  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes - Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
  • Calibration log stores ten most recent calibrations
  • Non volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with universal power adapter or optional AA batteries
  • Three year meter warranty

IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited.

Measure pH, mV, relative mV or ORP with temperature on channel one and measure dissolved oxygen in percent saturation or mg/L with temperature using RDO optical DO probes or polarographic DO probes on channel two.

Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values and calibration editing option that allows individual points to be fixed without a full recalibration. Calibrate dissolved oxygen using water-saturated air, air-saturated water, custom value (Winkler titration) and zero point options.

Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings.

 : Supplied with electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate.
The kit additionally includes: 8157BNUMD ROSS Ultra Triode refillable epoxy-body pH/ATC electrode, 083005MD Polarographic DO sensor with calibration sleeve, ROSS pH buffer and storage solution kit, DO sensor maintenance kit and BOD adapter, funnel and stirrer.

Specifications for Product Family

Specifications

  • Catalog No:
  • 665-0273
  • 665-0274
  • Model:
  • A216
  • A216
  • pH range:
  • –2,000...+20,000
  • –2,000...+20,000
  • pH resolution:
  • 0,1/0,01/0,001
  • 0,1/0,01/0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • pH calibration:
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • mV range:
  • ±2000,0
  • ±2000,0
  • mV resolution:
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • DO concentration:
  • 0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
  • 0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
  • DO concentration resolution:
  • 0,1/0,01 mg/L
  • 0,1/0,01 mg/L
  • DO saturation:
  • 0 - 600% (polarographic); 0 - 500% (RDO)
  • 0 - 600% (polarographic); 0 - 500% (RDO)
  • DO accuracy:
  • Polarographic: ±0,2 mg/L or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/L (up to 8 mg/L), ±0,2 mg/L (8 - 20 mg/L), ±10% of reading (up to 50 mg/L)
  • Polarographic: ±0,2 mg/L or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/L (up to 8 mg/L), ±0,2 mg/L (8 - 20 mg/L), ±10% of reading (up to 50 mg/L)
  • DO calibration:
  • Water-saturated air, air saturated water, manual (Winkler) and zero point
  • Water-saturated air, air saturated water, manual (Winkler) and zero point
  • DO barometric pressure range:
  • Automatic or manual, 450 - 850 mm Hg
  • Automatic or manual, 450 - 850 mm Hg
  • DO range:
  • 0,00 - 90,00 mg/L
  • 0,00 - 90,00 mg/L
  • Temperature range:
  • –5,0...+50,0 °C
  • –5,0...+50,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Pressure range:
  • 400 - 850 mm Hg
  • 400 - 850 mm Hg
  • Display:
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Battery type:
  • 4 AA (sold separately)
  • 4 AA (sold separately)
  • Battery life:
  • 800 h
  • 800 h
  • Power supply:
  • AC adapter (included) or 4 AA batteries
  • AC adapter (included) or 4 AA batteries
  • Compliance:
  • CE, TUV 3-1, FCC Class A
  • CE, TUV 3-1, FCC Class A
  • Operating temperature range:
  • 5...45 °C (ambient)
  • 5...45 °C (ambient)
  • Data logging - log function:
  • Manual/Automatic with AUTO-READ™
  • Manual/Automatic with AUTO-READ™
  • Data logging capacity:
  • 2000 with date and time stamp
  • 2000 with date and time stamp
  • Memory:
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Languages:
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • IP rating:
  • IP 54
  • IP 54
  • Connections:
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
  • Outputs:
  • RS232, USB
  • RS232, USB
  • Input:
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
  • Alarm:
  • High/low limit alarm, calibration due alarm
  • High/low limit alarm, calibration due alarm
  • No. of channels:
  • 2
  • 2
  • Calibration:
  • pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
  • pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
  • Calibration points:
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • W×D×H:
  • 240×180×110 mm
  • 240×180×110 mm
  • Weight:
  • 0,9 kg
  • 0,9 kg

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • 665-0273
    Bench pH/DO meter Orion Star A216, instrument alone
    Bench pH/DO meter Orion Star A216, instrument alone
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 610,00 €
    1 610,00 €
    1 * 1 ST
       1 610,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    665-0273
    Model
    A216
    pH range
    –2,000...+20,000
    pH resolution
    0,1/0,01/0,001
    pH accuracy
    ±0,002
    pH calibration
    Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
    mV range
    ±2000,0
    mV resolution
    0,1
    mV accuracy
    ±0,2 mV or ±0,05% of reading, whichever is greater
    DO concentration
    0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
    DO concentration resolution
    0,1/0,01 mg/L
    DO saturation
    0 - 600% (polarographic); 0 - 500% (RDO)
    DO accuracy
    Polarographic: ±0,2 mg/L or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/L (up to 8 mg/L), ±0,2 mg/L (8 - 20 mg/L), ±10% of reading (up to 50 mg/L)
    DO calibration
    Water-saturated air, air saturated water, manual (Winkler) and zero point
    DO barometric pressure range
    Automatic or manual, 450 - 850 mm Hg
    DO range
    0,00 - 90,00 mg/L
    Temperature range
    –5,0...+50,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C
    Temperature compensation
    Automatic or manual
    Pressure range
    400 - 850 mm Hg
    Display
    Graphic LCD with backlight
    Battery type
    4 AA (sold separately)
    Battery life
    800 h
    Power supply
    AC adapter (included) or 4 AA batteries
    Compliance
    CE, TUV 3-1, FCC Class A
    Operating temperature range
    5...45 °C (ambient)
    Data logging - log function
    Manual/Automatic with AUTO-READ™
    Data logging capacity
    2000 with date and time stamp
    Memory
    Non-volatile memory preserves data log, calibration log and meter settings
    Languages
    English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
    IP rating
    IP 54
    Connections
    BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
    Outputs
    RS232, USB
    Input
    BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
    Alarm
    High/low limit alarm, calibration due alarm
    No. of channels
    2
    Calibration
    pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
    Calibration points
    1 point relative mV (RmV) mode, 1 point ORP mode
    W×D×H
    240×180×110 mm
    Weight
    0,9 kg
  • 665-0274
    Bench pH/DO meter Orion Star A216, kit incl. ROSS Ultra Triode pH/ATC probe and polarographic DO probe
    Bench pH/DO meter Orion Star A216, kit incl. ROSS Ultra Triode pH/ATC probe and polarographic DO probe
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 620,00 €
    2 620,00 €
    1 * 1 ST
       2 620,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    665-0274
    Model
    A216
    pH range
    –2,000...+20,000
    pH resolution
    0,1/0,01/0,001
    pH accuracy
    ±0,002
    pH calibration
    Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
    mV range
    ±2000,0
    mV resolution
    0,1
    mV accuracy
    ±0,2 mV or ±0,05% of reading, whichever is greater
    DO concentration
    0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
    DO concentration resolution
    0,1/0,01 mg/L
    DO saturation
    0 - 600% (polarographic); 0 - 500% (RDO)
    DO accuracy
    Polarographic: ±0,2 mg/L or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/L (up to 8 mg/L), ±0,2 mg/L (8 - 20 mg/L), ±10% of reading (up to 50 mg/L)
    DO calibration
    Water-saturated air, air saturated water, manual (Winkler) and zero point
    DO barometric pressure range
    Automatic or manual, 450 - 850 mm Hg
    DO range
    0,00 - 90,00 mg/L
    Temperature range
    –5,0...+50,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C
    Temperature compensation
    Automatic or manual
    Pressure range
    400 - 850 mm Hg
    Display
    Graphic LCD with backlight
    Battery type
    4 AA (sold separately)
    Battery life
    800 h
    Power supply
    AC adapter (included) or 4 AA batteries
    Compliance
    CE, TUV 3-1, FCC Class A
    Operating temperature range
    5...45 °C (ambient)
    Data logging - log function
    Manual/Automatic with AUTO-READ™
    Data logging capacity
    2000 with date and time stamp
    Memory
    Non-volatile memory preserves data log, calibration log and meter settings
    Languages
    English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
    IP rating
    IP 54
    Connections
    BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
    Outputs
    RS232, USB
    Input
    BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
    Alarm
    High/low limit alarm, calibration due alarm
    No. of channels
    2
    Calibration
    pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
    Calibration points
    1 point relative mV (RmV) mode, 1 point ORP mode
    W×D×H
    240×180×110 mm
    Weight
    0,9 kg
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Benchtop meter-attached electrode stand (electrode holder arm and head), Orion™ Star A series, 360° motion (without weighted base)
    Benchtop meter-attached electrode stand (electrode holder arm and head), Orion™ Star A series, 360° motion (without weighted base)
    ORIOSTARA-BEA
    Orion Star A110 + A210 series, Orion Versa Star benchtop meters
    Log in for availability
    1 * 1 ST - 163,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cable
    Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cable
    662-1140
    Log in for availability
    1 * 1 SET - 76,40 €

Product Information

  • Catalog #
    Availability
    Price
  • Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series meters
    Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series meters
    662-1547
    Log in for availability
    1 * 1 ST - 72,40 €

Product Information

  • Catalog #
    Availability
    Price
  • Orion™ Star™ A Series weighted base for use with meter-attached electrode stand
    Orion™ Star™ A Series weighted base for use with meter-attached electrode stand
    ORIOSTARA-HB
    Log in for availability
    1 * 1 ST - 56,30 €

Product Information

  • Catalog #
    Availability
    Price
  • RS232 cable, for connecting a meter to a PC or printer
    RS232 cable, for connecting a meter to a PC or printer
    ORIO1010053
    Log in for availability
    1 * 1 ST - 45,40 €

Product Information

  • Catalog #
    Availability
    Price
  • Cable adapter, for connecting the RS232 port of a meter to a USB port
    Cable adapter, for connecting the RS232 port of a meter to a USB port
    ORIO1010005
    Log in for availability
    1 * 1 ST - 131,00 €