Order Entry
Austria
ContactUsLinkComponent
pH/mV/°C meter, handheld, Orion Star™ A221
pH/mV/°C meter, handheld, Orion Star™ A221
Supplier:  Thermo Orion
Certificates
undefined
pH/mV/°C meter, handheld, Orion Star™ A221
Supplier:  Thermo Orion
Limited Time Sale
Select items below are currently on sale

Specifications

  • Catalog No:
  • ORIOSTARA2217
  • 662-1445
  • 662-1444
  • ORIOSTARA2216
  • Model:
  • A221
  • A221
  • A221
  • A221
  • pH range:
  • -2,000 – 20,000 pH
  • −2,000...+20,000
  • −2,000...+20,000
  • -2,000 – 20,000 pH
  • pH resolution:
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • pH calibration:
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • mV range:
  • ±2000,0
  • ±2000,0
  • ±2000,0
  • ±2000,0
  • mV resolution:
  • 0,1
  • 0,1
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • Temperature range:
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Automatic or manual
  • Display:
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Battery life:
  • 800 h.
  • 800 h.
  • 800 h.
  • 800 h.
  • Power supply:
  • 4 AA batteries (included) or universal AC adapter (sold separately)
  • AC adapter (not included) or 4 AA batteries
  • AC adapter (not included) or 4 AA batteries
  • 4 AA batteries (included) or universal AC adapter (sold separately)
  • Compliance:
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • Operating temperature range:
  • 5...45 °C (ambient)
  • 5...45 °C (ambient)
  • 5...45 °C (ambient)
  • 5...45 °C (ambient)
  • Data logging - log function:
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Data logging capacity:
  • 1000 with date and time stamp
  • 1000 with date and time stamp
  • 1000 with date and time stamp
  • 1000 with date and time stamp
  • Memory:
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Languages:
  • English, Spanish, French, Italian, German, and Chinese (Portuguese and Korean available with software update)
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German, and Chinese (Portuguese and Korean available with software update)
  • Connections:
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
  • BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
  • Outputs:
  • RS232, USB
  • RS232, USB
  • RS232, USB
  • RS232, USB
  • Alarm:
  • Calibration due alarm (fixed 12 hour)
  • Calibration due alarm (fixed 12 hour)
  • Calibration due alarm (fixed 12 hour)
  • Calibration due alarm (fixed 12 hour)
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • Calibration:
  • pH, relative mV (RmV), ORP and temperature
  • pH, relative mV (RmV), ORP and temperature
  • pH, relative mV (RmV), ORP and temperature
  • pH, relative mV (RmV), ORP and temperature
  • Calibration points:
  • 1 (relative mV), 1 (ORP)
  • 1 - 5
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 - 5
  • W×D×H:
  • 240×100×65 mm
  • 240×100×65 mm
  • 240×100×65 mm
  • 240×100×65 mm
  • Weight:
  • 0,5 kg
  • 0,5 kg
  • 0,5 kg
  • 0,5 kg

Specifications

Frequently Bought Together