Order Entry
Austria
ContactUsLinkComponent
pH/mV/ISE meter, handheld, Orion Star™ A324
pH/mV/ISE meter, handheld, Orion Star™ A324
Supplier:  Thermo Orion
Certificates
undefined
pH/mV/ISE meter, handheld, Orion Star™ A324
Supplier:  Thermo Orion

Specifications

  • Catalog No:
  • 665-0275
  • 665-0276
  • Model:
  • A324
  • A324
  • pH range:
  • –2,000...+20,000
  • –2,000...+20,000
  • pH resolution:
  • 0,1/0,01/0,001
  • 0,1/0,01/0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • pH calibration:
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • mV range:
  • ±2000,0
  • ±2000,0
  • mV resolution:
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • ISE range:
  • 0,0001 - 19900
  • 0,0001 - 19900
  • ISE resolution:
  • 0,0001 minimum, 1 to 3 significant digits (user selectable)
  • 0,0001 minimum, 1 to 3 significant digits (user selectable)
  • ISE accuracy:
  • ±0,2 or ±0,05% of reading
  • ±0,2 or ±0,05% of reading
  • ISE calibration:
  • Up to 5 points
  • Up to 5 points
  • ISE units:
  • ppm, M, mg/L, %, ppb, none
  • ppm, M, mg/L, %, ppb, none
  • Temperature range:
  • –5,0...+105,0 °C
  • –5,0...+105,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Battery type:
  • 4 AA (included)
  • 4 AA (included)
  • Battery life:
  • 800 h
  • 800 h
  • Power supply:
  • AC adapter or 4 AA batteries
  • AC adapter or 4 AA batteries
  • Compliance:
  • IP 67
  • IP 67
  • Data logging - log function:
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Data logging capacity:
  • 2000
  • 2000
  • Memory:
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Languages:
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • Connections:
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
  • BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
  • Outputs:
  • RS232, USB
  • RS232, USB
  • No. of channels:
  • 1
  • 1
  • Calibration:
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative
  • Calibration points:
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • W×D×H:
  • 240×100×65 mm
  • 240×100×65 mm
  • Weight:
  • 0,5 kg
  • 0,5 kg

Specifications

Frequently Bought Together