Order Entry
Austria
Orders LinkContactUsLinkComponent
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld

Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld

Supplier:   Thermo Orion
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld
Supplier:  Thermo Orion
Certificates
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld
Orion Star™ A329 Multi-Parameter Meter (pH/ISE/Conductivity/DO), Handheld

2 Options Available Below

Product Details & Documents

This rugged, waterproof portable meter is designed for a wide range of pH, ion concentration, conductivity, dissolved oxygen and temperature testing and field applications. It can be used in the most demanding locations thanks to its IP67-rated housing. The meter offers three measuring channels allowing users to measure pH, conductivity and DO simultaneously or view each channel separately.

  • Up to five point pH calibrations with calibration editing option that allows individual points to be fixed without a full recalibration
  • Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
  • Calibration log stores ten most recent calibrations
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections

Measure pH, mV, relative mV or ORP with temperature on channel one and measure dissolved oxygen in percent saturation or mg/L with temperature using RDO optical DO probes or polarographic DO probes on channel two.

Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings. Transfer single, range or all data points to printer or computer.

Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate. Use linear, non linear, EP (USP) or off conductivity temperature compensation with 5, 10, 15, 20 or 25 °C reference temperatures. 3-years replacement warranty.

Certifications: TUV 3-1, FCC Class A.

Delivery information: Supplied with four with 4 AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate.
The kit additionally includes: 8107UWMMD ROSS Ultra Triode gel filled epoxy-body pH/ATC electrode (3 m cable), 013010MD DuraProbe 4-cell (K=0,475) epoxy-body conductivity sensor (3 m cable), 087010MD RDO optical DO sensor (3 m cable), calibration sleeve and guard, Orion pH 4/7/10 buffer pouches (10 packs each), ROSS electrode storage solution (475 ml), Orion rinse solution pouches (10 packs), Orion 1413 µS conductivity standard pouches (10 packs), Orion Star A series portable meter armor and Orion Star A series hard-sided field case.

Specifications for Product Family

Specifications

  • Catalog No:
  • 665-0281
  • 665-0282
  • Model:
  • A329
  • A329
  • pH range:
  • –2,000...+20,000
  • –2,000...+20,000
  • pH resolution:
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • pH calibration:
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • mV range:
  • ±2000,0
  • ±2000,0
  • mV resolution:
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or 0,05%
  • ±0,2 mV or 0,05%
  • ISE range:
  • 0,0001 - 19900
  • 0,0001 - 19900
  • ISE resolution:
  • 0,0001 minimum, 1 to 3 significant digits (user selectable)
  • 0,0001 minimum, 1 to 3 significant digits (user selectable)
  • ISE accuracy:
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • ±0,2 mV or ±0,05% of reading, whichever is greater
  • ISE calibration:
  • Up to 5 points
  • Up to 5 points
  • ISE units:
  • ppm, M, mg/l, %, ppb, none
  • ppm, M, mg/l, %, ppb, none
  • Conductivity range:
  • 0,001 µS/cm - 3000 mS/cm
  • 0,001 µS/cm - 3000 mS/cm
  • Conductivity resolution:
  • 0,001 μS minimum, auto ranging, up to 4 significant digits
  • 0,001 μS minimum, auto ranging, up to 4 significant digits
  • Conductivity accuracy:
  • ±0,5% of reading ±1 digit
  • ±0,5% of reading ±1 digit
  • Conductivity cell constant:
  • 0,001 - 199,9 cm⁻¹
  • 0,001 - 199,9 cm⁻¹
  • Conductivity calibration:
  • Up to 5 points
  • Up to 5 points
  • Conductivity reference temperature:
  • 5/10/15/20/25 °C
  • 5/10/15/20/25 °C
  • Resistivity range:
  • 2 Ω/cm - 100 MΩ/cm
  • 2 Ω/cm - 100 MΩ/cm
  • Resistivity accuracy:
  • 0,5% of reading ±1 digit
  • 0,5% of reading ±1 digit
  • Resistivity resolution:
  • 1 ohm or 0,1 meg-ohm, auto ranging
  • 1 ohm or 0,1 meg-ohm, auto ranging
  • Salinity range:
  • 0,06 - 80,00 psu / 0,05 - 42,00 ppt
  • 0,06 - 80,00 psu / 0,05 - 42,00 ppt
  • Salinity resolution:
  • 0,01 psu or 0,01 ppt minimum, auto ranging
  • 0,01 psu or 0,01 ppt minimum, auto ranging
  • Salinity accuracy:
  • 0,5% of reading ±1 digit
  • 0,5% of reading ±1 digit
  • TDS range:
  • 0,001 - 200,0 ppm
  • 0,001 - 200,0 ppm
  • TDS resolution:
  • 0,001 ppm minimum, auto ranging, up to 4 significant digits
  • 0,001 ppm minimum, auto ranging, up to 4 significant digits
  • TDS accuracy:
  • 0,5% of reading ±1 digit
  • 0,5% of reading ±1 digit
  • TDS conversion factor:
  • Linear (0,02 - 9,99)
  • Linear (0,02 - 9,99)
  • DO concentration:
  • 0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
  • 0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
  • DO concentration resolution:
  • 0,1 / 0,01 mg/L
  • 0,1 / 0,01 mg/L
  • DO saturation:
  • 0 - 600 % (polarographic); 0 - 500 % (RDO)
  • 0 - 600 % (polarographic); 0 - 500 % (RDO)
  • DO accuracy:
  • Polarographic: ±0,2 mg/l or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/l (up to 8 mg/), ±0,2 mg/l (8 - 20 mg/l), ±10% of reading (up to 50 mg/l)
  • Polarographic: ±0,2 mg/l or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/l (up to 8 mg/), ±0,2 mg/l (8 - 20 mg/l), ±10% of reading (up to 50 mg/l)
  • DO calibration:
  • Water-saturated air, air saturated water, manual (Winkler) and zero point
  • Water-saturated air, air saturated water, manual (Winkler) and zero point
  • DO barometric pressure range:
  • Automatic or manual, 450 - 850 mmHg
  • Automatic or manual, 450 - 850 mmHg
  • Temperature range:
  • –5,0...+105,0 °C (–5,0...+50,0 °C with DO)
  • –5,0...+105,0 °C (–5,0...+50,0 °C with DO)
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • Temperature calibration:
  • 1 point temperature offset calibration
  • 1 point temperature offset calibration
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Pressure range:
  • 400 - 850 mm Hg
  • 400 - 850 mm Hg
  • Pressure resolution:
  • 1 mm Hg
  • 1 mm Hg
  • Display:
  • Graphic LCD with backlight
  • Graphic LCD with backlight
  • Battery type:
  • 4 AA (included)
  • 4 AA (included)
  • Battery life:
  • 800 hours
  • 800 hours
  • Power supply:
  • AC adapter or 4 AA batteries
  • AC adapter or 4 AA batteries
  • Compliance:
  • IP 67
  • IP 67
  • Data logging - log function:
  • Manual / Automatic with AUTO-READ™
  • Manual / Automatic with AUTO-READ™
  • Data logging capacity:
  • 5000
  • 5000
  • Memory:
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Non-volatile memory preserves data log, calibration log and meter settings
  • Languages:
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
  • IP rating:
  • IP 67
  • IP 67
  • Connections:
  • BNC, pin tip, 8- and 9-pin mini-DIN
  • BNC, pin tip, 8- and 9-pin mini-DIN
  • Outputs:
  • RS232, USB
  • RS232, USB
  • Input:
  • 3 measuring channels
  • 3 measuring channels
  • Alarm:
  • High/low limit alarm, calibration due alarm
  • High/low limit alarm, calibration due alarm
  • No. of channels:
  • 3
  • 3
  • Calibration:
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
  • pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
  • Calibration points:
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • 1 point relative mV (RmV) mode, 1 point ORP mode
  • W×D×H:
  • 60×105×240 mm
  • 60×105×240 mm
  • Weight:
  • 0,5 kg
  • 0,5 kg

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • 665-0281
    Handheld pH/ISE/conductivity/DO meter Orion Star A329, instrument alone
    Handheld pH/ISE/conductivity/DO meter Orion Star A329, instrument alone
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 030,00 €
    2 030,00 €
    1 * 1 ST
       2 030,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    665-0281
    Model
    A329
    pH range
    –2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    pH calibration
    Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
    mV range
    ±2000,0
    mV resolution
    0,1
    mV accuracy
    ±0,2 mV or 0,05%
    ISE range
    0,0001 - 19900
    ISE resolution
    0,0001 minimum, 1 to 3 significant digits (user selectable)
    ISE accuracy
    ±0,2 mV or ±0,05% of reading, whichever is greater
    ISE calibration
    Up to 5 points
    ISE units
    ppm, M, mg/l, %, ppb, none
    Conductivity range
    0,001 µS/cm - 3000 mS/cm
    Conductivity resolution
    0,001 μS minimum, auto ranging, up to 4 significant digits
    Conductivity accuracy
    ±0,5% of reading ±1 digit
    Conductivity cell constant
    0,001 - 199,9 cm⁻¹
    Conductivity calibration
    Up to 5 points
    Conductivity reference temperature
    5/10/15/20/25 °C
    Resistivity range
    2 Ω/cm - 100 MΩ/cm
    Resistivity accuracy
    0,5% of reading ±1 digit
    Resistivity resolution
    1 ohm or 0,1 meg-ohm, auto ranging
    Salinity range
    0,06 - 80,00 psu / 0,05 - 42,00 ppt
    Salinity resolution
    0,01 psu or 0,01 ppt minimum, auto ranging
    Salinity accuracy
    0,5% of reading ±1 digit
    TDS range
    0,001 - 200,0 ppm
    TDS resolution
    0,001 ppm minimum, auto ranging, up to 4 significant digits
    TDS accuracy
    0,5% of reading ±1 digit
    TDS conversion factor
    Linear (0,02 - 9,99)
    DO concentration
    0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
    DO concentration resolution
    0,1 / 0,01 mg/L
    DO saturation
    0 - 600 % (polarographic); 0 - 500 % (RDO)
    DO accuracy
    Polarographic: ±0,2 mg/l or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/l (up to 8 mg/), ±0,2 mg/l (8 - 20 mg/l), ±10% of reading (up to 50 mg/l)
    DO calibration
    Water-saturated air, air saturated water, manual (Winkler) and zero point
    DO barometric pressure range
    Automatic or manual, 450 - 850 mmHg
    Temperature range
    –5,0...+105,0 °C (–5,0...+50,0 °C with DO)
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C
    Temperature calibration
    1 point temperature offset calibration
    Temperature compensation
    Automatic or manual
    Pressure range
    400 - 850 mm Hg
    Pressure resolution
    1 mm Hg
    Display
    Graphic LCD with backlight
    Battery type
    4 AA (included)
    Battery life
    800 hours
    Power supply
    AC adapter or 4 AA batteries
    Compliance
    IP 67
    Data logging - log function
    Manual / Automatic with AUTO-READ™
    Data logging capacity
    5000
    Memory
    Non-volatile memory preserves data log, calibration log and meter settings
    Languages
    English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
    IP rating
    IP 67
    Connections
    BNC, pin tip, 8- and 9-pin mini-DIN
    Outputs
    RS232, USB
    Input
    3 measuring channels
    Alarm
    High/low limit alarm, calibration due alarm
    No. of channels
    3
    Calibration
    pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
    Calibration points
    1 point relative mV (RmV) mode, 1 point ORP mode
    W×D×H
    60×105×240 mm
    Weight
    0,5 kg
  • 665-0282
    Handheld pH/ISE/conductivity/DO meter Orion Star A329, kit incl. pH electrode, conductivity cell and RDO optical sensor
    Handheld pH/ISE/conductivity/DO meter Orion Star A329, kit incl. pH electrode, conductivity cell and RDO optical sensor
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 060,00 €
    3 060,00 €
    1 * 1 ST
       3 060,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    665-0282
    Model
    A329
    pH range
    –2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    pH calibration
    Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
    mV range
    ±2000,0
    mV resolution
    0,1
    mV accuracy
    ±0,2 mV or 0,05%
    ISE range
    0,0001 - 19900
    ISE resolution
    0,0001 minimum, 1 to 3 significant digits (user selectable)
    ISE accuracy
    ±0,2 mV or ±0,05% of reading, whichever is greater
    ISE calibration
    Up to 5 points
    ISE units
    ppm, M, mg/l, %, ppb, none
    Conductivity range
    0,001 µS/cm - 3000 mS/cm
    Conductivity resolution
    0,001 μS minimum, auto ranging, up to 4 significant digits
    Conductivity accuracy
    ±0,5% of reading ±1 digit
    Conductivity cell constant
    0,001 - 199,9 cm⁻¹
    Conductivity calibration
    Up to 5 points
    Conductivity reference temperature
    5/10/15/20/25 °C
    Resistivity range
    2 Ω/cm - 100 MΩ/cm
    Resistivity accuracy
    0,5% of reading ±1 digit
    Resistivity resolution
    1 ohm or 0,1 meg-ohm, auto ranging
    Salinity range
    0,06 - 80,00 psu / 0,05 - 42,00 ppt
    Salinity resolution
    0,01 psu or 0,01 ppt minimum, auto ranging
    Salinity accuracy
    0,5% of reading ±1 digit
    TDS range
    0,001 - 200,0 ppm
    TDS resolution
    0,001 ppm minimum, auto ranging, up to 4 significant digits
    TDS accuracy
    0,5% of reading ±1 digit
    TDS conversion factor
    Linear (0,02 - 9,99)
    DO concentration
    0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
    DO concentration resolution
    0,1 / 0,01 mg/L
    DO saturation
    0 - 600 % (polarographic); 0 - 500 % (RDO)
    DO accuracy
    Polarographic: ±0,2 mg/l or ±2% of reading, whichever is greater
    RDO: ±0,1 mg/l (up to 8 mg/), ±0,2 mg/l (8 - 20 mg/l), ±10% of reading (up to 50 mg/l)
    DO calibration
    Water-saturated air, air saturated water, manual (Winkler) and zero point
    DO barometric pressure range
    Automatic or manual, 450 - 850 mmHg
    Temperature range
    –5,0...+105,0 °C (–5,0...+50,0 °C with DO)
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C
    Temperature calibration
    1 point temperature offset calibration
    Temperature compensation
    Automatic or manual
    Pressure range
    400 - 850 mm Hg
    Pressure resolution
    1 mm Hg
    Display
    Graphic LCD with backlight
    Battery type
    4 AA (included)
    Battery life
    800 hours
    Power supply
    AC adapter or 4 AA batteries
    Compliance
    IP 67
    Data logging - log function
    Manual / Automatic with AUTO-READ™
    Data logging capacity
    5000
    Memory
    Non-volatile memory preserves data log, calibration log and meter settings
    Languages
    English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
    IP rating
    IP 67
    Connections
    BNC, pin tip, 8- and 9-pin mini-DIN
    Outputs
    RS232, USB
    Input
    3 measuring channels
    Alarm
    High/low limit alarm, calibration due alarm
    No. of channels
    3
    Calibration
    pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
    Calibration points
    1 point relative mV (RmV) mode, 1 point ORP mode
    W×D×H
    60×105×240 mm
    Weight
    0,5 kg
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cable
    Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cable
    662-1140
    Thermo Scientific™ Orion™ Versa Star meters, Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus meters
    Log in for availability
    1 * 1 SET - 76,40 €

Product Information

  • Catalog #
    Availability
    Price
  • Orion™ Star™ A Series portable meter pH electrode holder, can be attached to the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeve
    Orion™ Star™ A Series portable meter pH electrode holder, can be attached to the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeve
    ORIOSTARA-ESPH
    Log in for availability
    1 * 1 ST - 29,50 €

Product Information

  • Catalog #
    Availability
    Price
  • Hard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve
    Hard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve
    ORIOSTARA-CS
    Log in for availability
    1 * 1 ST - 144,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series meters
    Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series meters
    662-1547
    Log in for availability
    1 * 1 ST - 72,40 €

Product Information

  • Catalog #
    Availability
    Price
  • Orion™ Star A Series portable meter conductivity and DO probe holder, can be attached to either side of the portable meter armor and holds 12/15 mm Ø conductivity probes and a dissolved oxygen probe when placed in its calibration sleeve
    Orion™ Star A Series portable meter conductivity and DO probe holder, can be attached to either side of the portable meter armor and holds 12/15 mm Ø conductivity probes and a dissolved oxygen probe when placed in its calibration sleeve
    ORIOSTARA-ESCD
    Log in for availability
    1 * 1 ST - 29,50 €

Product Information

  • Catalog #
    Availability
    Price
  • Protective armor for portable meters, incl. hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, conductivity and dissolved oxygen sensor holder
    Protective armor for portable meters, incl. hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, conductivity and dissolved oxygen sensor holder
    ORIOSTARA-AR
    Log in for availability
    1 * 1 ST - 102,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Soft-sided field case, incl. interior dividers and exterior pockets to safely organise the meter and accessories in the field
    Soft-sided field case, incl. interior dividers and exterior pockets to safely organise the meter and accessories in the field
    ORIO1210005
    Log in for availability
    1 * 1 ST - 161,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • RS232 cable, for connecting a meter to a PC or printer
    RS232 cable, for connecting a meter to a PC or printer
    ORIO1010053
    Orion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series), Orion™ Dual Star benchtop meters, Orion 3-Star, 4-Star and 5-Star meters
    Log in for availability
    1 * 1 ST - 45,40 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Cable adapter, for connecting the RS232 port of a meter to a USB port
    Cable adapter, for connecting the RS232 port of a meter to a USB port
    ORIO1010005
    Orion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series) and Orion™ Star and Star Plus meters
    Log in for availability
    1 * 1 ST - 131,00 €