Order Entry
Austria
ContactUsLinkComponent
pH/mV/°C meters, handheld, Orion™ Star A121
pH/mV/°C meters, handheld, Orion™ Star A121
Supplier:  Thermo Orion
Certificates
undefined
pH/mV/°C meters, handheld, Orion™ Star A121
Supplier:  Thermo Orion
Limited Time Sale
Select items below are currently on sale

Specifications

  • Catalog No:
  • 662-1440
  • 662-1441
  • Model:
  • A121
  • A121
  • pH range:
  • −2,00...+16,00
  • −2,00...+16,00
  • pH resolution:
  • 0,1/0,01
  • 0,1/0,01
  • pH accuracy:
  • ±0,01
  • ±0,01
  • pH calibration:
  • Up to 3 points; Automatic buffer recognition (USA/NIST and DIN)
  • Up to 3 points; Automatic buffer recognition (USA/NIST and DIN)
  • mV range:
  • ±1600,0/±1999,9
  • ±1600,0/±1999,9
  • mV resolution:
  • 0,1
  • 0,1
  • mV accuracy:
  • ±0,2 mV or ±0,05% of reading whichever is greater
  • ±0,2 mV or ±0,05% of reading whichever is greater
  • Temperature range:
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • Temperature compensation:
  • Automatic or manual
  • Automatic or manual
  • Display:
  • LCD
  • LCD
  • Battery type:
  • 4×AA
  • 4×AA
  • Power supply:
  • AC adapter (not included) or 4 AA batteries
  • AC adapter (not included) or 4 AA batteries
  • Compliance:
  • IP 67
  • IP 67
  • Operating temperature range:
  • 5...45 °C (ambient)
  • 5...45 °C (ambient)
  • Data logging - log function:
  • Manual/Automatic with AUTO-READ™
  • Manual/Automatic with AUTO-READ™
  • Data logging capacity:
  • 50
  • 50
  • Memory:
  • Non-volatile memory preserves log and settings
  • Non-volatile memory preserves log and settings
  • Languages:
  • English
  • English
  • Connections:
  • BNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
  • BNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
  • No. of channels:
  • 1
  • 1
  • Calibration points:
  • 1 - 3
  • 1 - 3
  • W×D×H:
  • 100×240×65 mm
  • 100×240×65 mm
  • Weight:
  • 0,5 kg
  • 0,5 kg

Specifications

Electrodes

Product Information