Order Entry
Austria
ContactUsLinkComponent
pH / LogR measurement module for Orion™ Versa Star Pro™ meters
pH / LogR measurement module for Orion™ Versa Star Pro™ meters
Catalog # ORIOVSTAR-LR
Supplier:  Thermo Orion
undefined
pH / LogR measurement module for Orion™ Versa Star Pro™ meters
Catalog # ORIOVSTAR-LR
Supplier:  Thermo Orion

Specifications

  • Calibration:
    pH with calibration editing option, relative mV (RmV), ORP, LogR temperature and temperature
  • Description:
    pH/LogR/mV/RmV/ORP/temperature module for Orion™ Versa Star Pro™ meters (LogR technology uses the glass-bulb of most pH electrodes to measure both pH and temperature at the same location)
  • Seq:
    03
  • Temperature accuracy:
    ±0,1 °C
  • Alarm:
    Set point alarm, high/low limit alarm, calibration due alarm
  • Calibration points:
    1 to 6
  • W×D×H:
    120×60×95 mm
  • mV accuracy:
    ±0,2 mV or ±0,05% of reading whichever is greater
  • pH accuracy:
    ±0,002 pH
  • Pk:
    1

Specifications

About this item

This measurement module can be used to customise the measurement capabilities of the Orion™ Versa Star Pro™ meters. It adds pH, mV, ORP and LogR temperature measurement capabilities to the meter. The module offers LogR temperature technology that uses the glass-bulb of most pH electrodes to measure both pH and temperature at the same location – ideal for small sample volumes and samples that are difficult to clean.

  • Each Orion™ Versa Star Pro meter has four channels that can be customised with interchangeable measurement modules
  • Measurement modules are interchangeable
  • Up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customise measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading

Range (pH): −2,000 to +20,000
Range (mV): ±2000,0
Range (Temperature): −5 to +105 °C

3-year replacement warranty.