Order Entry
Austria
ContactUsLinkComponent
 
pH measurement module for Orion™ Versa Star Pro™ meters
  ORIOVSTAR-PH
undefined
pH measurement module for Orion™ Versa Star Pro™ meters
  ORIOVSTAR-PH

 

  • Description:
    pH/mV/temperature module for Orion™ Versa Star Pro™ meters
  • Seq:
    01
  • Pk:
    1

 

 

This measurement module can be used to customise the measurement capabilities of the Orion™ Versa Star Pro™ meters. It adds pH, mV, ORP and temperature measurement capabilities to the meter.

  • Each Orion™ Versa Star Pro meter has four channels that can be customised with interchangeable measurement modules
  • Measurement modules are interchangeable
  • Up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customise measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading

Range (pH): −2,000 to +20,000
Range (mV): ±2000,0
Range (Temperature): −5,0 to +105,0 °C

3-year replacement warranty.