ContactUsLinkComponent
LED illumination systems, HB 100, for microscopes
LED illumination systems, HB 100, for microscopes
Catalog # ZEIS4230119901000
Supplier:  ZEISS
undefined
LED illumination systems, HB 100, for microscopes
Catalog # ZEIS4230119901000
Supplier:  ZEISS
Specifications
  • Description:
    HBO 100 illuminator
  • Pk:
    1
  • For:
    Microscopes
Specifications
About this item
HBO 100 illuminator, self-adjusting with lamp mount and collector, including heat-reflecting filter and 2 m connection cable. Automatic lamp alignment.
  • Automatic lamp alignment